Learn More
Ace Glass, Inc. Adapter, 105°, 45/50 outer top and inner bottom
SDP
Catalog No. 50971698
Encompass_Preferred
Angle adapter with 105 degree 45/50 standard taper outer top joint and 45/50 standard taper inner bottom joint.
Catalog No. 50-971-698 Supplier Ace Glass, Inc. Supplier No. 507545
Angle adapter with 105 degree 45/50 standard taper outer top joint and 45/50 standard taper inner bottom joint.