Learn More
Ace Glass, Inc. 45.5L (12 gallon) solution bottle, GL120 threaded neck, no cap
SDP
Catalog No. 50971551
Encompass
45.5L solution bottle with 106mm I.D. opening and GL120 threaded neck. Bottle shape is similar in design to a carboy style. Cap not included, see ACE 7622-155.
Catalog No. 50-971-551 Supplier Ace Glass, Inc. Supplier No. 404845
45.5L solution bottle with 106mm I.D. opening and GL120 threaded neck. Bottle shape is similar in design to a carboy style. Cap not included, see ACE 7622-155.