Learn More
Electron Microscopy Sciences 45 degree Pin Stub Holder, 3.2 mm x 8mm Pin
SDP
Catalog No. 501901520
Encompass
This 45 degree pin stub holder allows for a higher SE signal without having to tilt the specimen stage and it accommodates any standard 12.7 mm (1/2") pin stubs with 3.2 mm (1/8") pin.. Grooved, machined aluminum with stainless steel Allen set screw.
Type: 3.2 mm x 8 mm Pin
Catalog No. 50-190-1520 Supplier Electron Microscopy Sciences Supplier No. 7595245
This 45 degree pin stub holder allows for a higher SE signal without having to tilt the specimen stage and it accommodates any standard 12.7 mm (1/2") pin stubs with 3.2 mm (1/8") pin.. Grooved, machined aluminum with stainless steel Allen set screw.
Type: 3.2 mm x 8 mm Pin