Learn More
Roboz Surgical 45 DEGREE ANGLE MICRO DISSECTI
SDP
Catalog No. NC9976442
Encompass
Dumont Tweezers; Pattern #SS, Long; Narrow, Delicate; 45 Degree Angle; .10 X .06mm Tip Size; Stainless Steel; 135mm Length; Material: Inox; Biologie Tips
Catalog No. NC9976442 Supplier Roboz Surgical Supplier No. RS4941
Dumont Tweezers; Pattern #SS, Long; Narrow, Delicate; 45 Degree Angle; .10 X .06mm Tip Size; Stainless Steel; 135mm Length; Material: Inox; Biologie Tips