Learn More
Grainger 45 Elbow SS 2 in Socket x Push Fit
Catalog No. 502748915
Encompass_Preferred
The 2" stainless steel 45 degree elbow from Grainger has a socket end and a push-fit end. It is made from 304 stainless steel for corrosion resistance. The maximum pressure rating is 300 PSI at 70 degrees F. The socket end accepts 2" Schedule 40 pipe. The push-fit end connects to 2" OD tube. This elbow allows you to make 45 degree turns in stainless steel tubing runs. It can be used in industrial compressed air, water, steam, gas, and chemical applications.
Catalog No. 50-274-8915 Supplier Grainger Supplier No. 14N965
The 2" stainless steel 45 degree elbow from Grainger has a socket end and a push-fit end. It is made from 304 stainless steel for corrosion resistance. The maximum pressure rating is 300 PSI at 70 degrees F. The socket end accepts 2" Schedule 40 pipe. The push-fit end connects to 2" OD tube. This elbow allows you to make 45 degree turns in stainless steel tubing runs. It can be used in industrial compressed air, water, steam, gas, and chemical applications.