Learn More
Grainger 45 Elbow SS 3 in Socket x Push Fit
Catalog No. 502748916
Encompass_Preferred
The BLUCHER 45-Degree Elbow is made of stainless steel for durability and corrosion resistance. It has a 45-degree angle and is designed for use in commercial food processing facilities to transport liquids. The elbow has a clamp connection end that is 2 inches in diameter, allowing it to connect to 2 inch stainless steel tubing. It can withstand temperatures up to 250°F and pressures up to 232 PSI. The elbow complies with 3-A sanitary standards.
Catalog No. 50-274-8916 Supplier Grainger Supplier No. 14N966
The BLUCHER 45-Degree Elbow is made of stainless steel for durability and corrosion resistance. It has a 45-degree angle and is designed for use in commercial food processing facilities to transport liquids. The elbow has a clamp connection end that is 2 inches in diameter, allowing it to connect to 2 inch stainless steel tubing. It can withstand temperatures up to 250°F and pressures up to 232 PSI. The elbow complies with 3-A sanitary standards.