Learn More
Grainger 45 Elbow SS 4 in Socket x Push Fit
Catalog No. 502748917
Encompass_Preferred
The Polaris Insulated Multitap Connector is an electrical connector that allows multiple wires to be connected to a single point. It is rated for 600 volts and can accommodate wire sizes from 14-10 AWG. The connector has 4 ports and is made from thermoplastic polyester with nickel-plated brass terminals. It is UL listed and CSA certified. The multitap is insulated to help prevent shorts and is ideal for splicing and extending circuits. This connector can be installed by stripping the wires, inserting them into the ports, and tightening the screws to secure the wires. It is manufactured by Polaris in the United States.
Catalog No. 50-274-8917 Supplier Grainger Supplier No. 14N967
The Polaris Insulated Multitap Connector is an electrical connector that allows multiple wires to be connected to a single point. It is rated for 600 volts and can accommodate wire sizes from 14-10 AWG. The connector has 4 ports and is made from thermoplastic polyester with nickel-plated brass terminals. It is UL listed and CSA certified. The multitap is insulated to help prevent shorts and is ideal for splicing and extending circuits. This connector can be installed by stripping the wires, inserting them into the ports, and tightening the screws to secure the wires. It is manufactured by Polaris in the United States.