Learn More
Sigma Aldrich Fine Chemicals Biosciences 45-Methylenedioxy-12-pheny10MG
Catalog No. 502898715
Encompass
4 5-Methylenedioxy-1 2-phenylenediamine dihydrochloride is a fluorescence tag for Aº-keto acids. It is also required for the derivatization of sialic acids in presence of dilute sulfuric acid. The method allows determination of sialic acids.
Catalog No. 50-289-8715 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 6680710MG
4 5-Methylenedioxy-1 2-phenylenediamine dihydrochloride is a fluorescence tag for Aº-keto acids. It is also required for the derivatization of sialic acids in presence of dilute sulfuric acid. The method allows determination of sialic acids.