Learn More
Ted Pella Inc 45º SEM Mount, 10/pack
SDP
Catalog No. NC3244329
Encompass
45º SEM Mount Ø12.7mm for FEI, TESCAN, ZEISS (also for Philips, LEO, Cambridge, AMRAY, Leica, CamScan, ETEC) SEMs. Aluminum, grooved edge, 7.8mm pin height.
Catalog No. NC3244329 Supplier Ted Pella Inc Supplier No. 16101
45º SEM Mount Ø12.7mm for FEI, TESCAN, ZEISS (also for Philips, LEO, Cambridge, AMRAY, Leica, CamScan, ETEC) SEMs. Aluminum, grooved edge, 7.8mm pin height.