Learn More
Chemglass Life Sciences Cap, #45, With Hole, Rodaviss
SDP
Supplier:  Chemglass Life Sciences CGB18206
Encompass_Preferred
Cap, #45, With Hole, Rodaviss. Threaded cap fits CG-172 outer joint, Rodaviss. When used with CG-305 Viton o-rings, provides for greaseless high vacuum seal.
Catalog No. 50-153-1931