Learn More
Chemglass Life Sciences Silicone Sealing Ring, fits GL-45 Cap, fits tubing O.D. 25.0mm to 27.0mm
SDP
Catalog No. CG19705
Encompass_Preferred
Silicone Sealing Ring, fits GL-45 Cap, fits tubing O.D. 25.0mm to 27.0mm. Seal is made from silicone with a vulcanized PTFE washer having an aperture to fit the listed tubing size. For use with CG-195 cap.
Catalog No. CG19705 Supplier Chemglass Life Sciences Supplier No. CG19705
Silicone Sealing Ring, fits GL-45 Cap, fits tubing O.D. 25.0mm to 27.0mm. Seal is made from silicone with a vulcanized PTFE washer having an aperture to fit the listed tubing size. For use with CG-195 cap.