Learn More
Grainger 45 Wye 316 SS 3 in Pipe Size 29 PSI
Catalog No. 502748735
Encompass_Preferred
The Blucher 45 Wye Stainless Steel is a 45 degree wye fitting made from 304 stainless steel. It has a 2 inch diameter and is designed for use in drain, waste and vent piping applications. The fitting allows a 45 degree change of direction in the pipe run. It is made in the USA and is lead free. The fitting has a maximum pressure rating of 300 psi at 70 degrees Fahrenheit. It conforms to ASTM A312 standards. The fitting would be useful for creating offsets and directional changes in stainless steel drain piping systems.
Catalog No. 50-274-8735 Supplier Grainger Supplier No. 14N989
The Blucher 45 Wye Stainless Steel is a 45 degree wye fitting made from 304 stainless steel. It has a 2 inch diameter and is designed for use in drain, waste and vent piping applications. The fitting allows a 45 degree change of direction in the pipe run. It is made in the USA and is lead free. The fitting has a maximum pressure rating of 300 psi at 70 degrees Fahrenheit. It conforms to ASTM A312 standards. The fitting would be useful for creating offsets and directional changes in stainless steel drain piping systems.