Learn More
Grainger 45 Wye 316 SS 4 in Pipe Size Socket
Catalog No. 502749539
Encompass_Preferred
The Grainger Approved 45° Wye is a PVC pipe fitting that creates a 45 degree branch connection. It is made of PVC Schedule 40 and has a maximum temperature rating of 140°F. The fitting accepts 3" PVC pipe and has a pressure rating of 480 psi at 73°F. It can be used for drain, waste and vent applications. The dimensions are 10-1/2"L x 10-1/2"W x 8-1/2"H and it weighs 2 lbs. This fitting provides a sturdy 45 degree PVC branch connection for 3" diameter PVC piping systems.
Catalog No. 50-274-9539 Supplier Grainger Supplier No. 14N991
The Grainger Approved 45° Wye is a PVC pipe fitting that creates a 45 degree branch connection. It is made of PVC Schedule 40 and has a maximum temperature rating of 140°F. The fitting accepts 3" PVC pipe and has a pressure rating of 480 psi at 73°F. It can be used for drain, waste and vent applications. The dimensions are 10-1/2"L x 10-1/2"W x 8-1/2"H and it weighs 2 lbs. This fitting provides a sturdy 45 degree PVC branch connection for 3" diameter PVC piping systems.