Learn More
Grainger 45 Wye 316 SS 4 x 2 x 4 in Pipe Size
Catalog No. 502749768
Encompass_Preferred
The Grainger Approved 45° Wye is a PVC pipe fitting used to connect three pipes at a 45 degree angle. It has a maximum temperature rating of 140°F and is made of PVC Schedule 40 for use in DWV and industrial waste applications. This fitting has 3 socket ends that accept pipes with 3" outer diameters and is designed for use with solvent cements. It meets ASTM D2665 standards. The 45° wye allows for a smooth 45 degree change of direction in piping runs.
Catalog No. 50-274-9768 Supplier Grainger Supplier No. 14N988
The Grainger Approved 45° Wye is a PVC pipe fitting used to connect three pipes at a 45 degree angle. It has a maximum temperature rating of 140°F and is made of PVC Schedule 40 for use in DWV and industrial waste applications. This fitting has 3 socket ends that accept pipes with 3" outer diameters and is designed for use with solvent cements. It meets ASTM D2665 standards. The 45° wye allows for a smooth 45 degree change of direction in piping runs.