Learn More
Grainger 45 Wye 316 SS 4 x 3 x 4 in Pipe Size
Catalog No. 502751822
Encompass_Preferred
The Grainger Approved 45° Wye is a PVC pipe fitting used to connect pipes at a 45 degree angle. It has a maximum temperature rating of 140°F and maximum pressure rating of 100 PSI. The fitting has solvent weld sockets and is made of PVC Schedule 40. It is compliant with ASTM D2466 standards. The fitting has a nominal pipe size of 1-1/2" and is gray in color. The 45° wye allows for a smooth 45 degree change of direction in a PVC piping system.
Catalog No. 50-275-1822 Supplier Grainger Supplier No. 14N990
The Grainger Approved 45° Wye is a PVC pipe fitting used to connect pipes at a 45 degree angle. It has a maximum temperature rating of 140°F and maximum pressure rating of 100 PSI. The fitting has solvent weld sockets and is made of PVC Schedule 40. It is compliant with ASTM D2466 standards. The fitting has a nominal pipe size of 1-1/2" and is gray in color. The 45° wye allows for a smooth 45 degree change of direction in a PVC piping system.