Learn More
Surmodics Inc 450 nm Liquid Nova-Stop Solution for TMB Microwell Substrates 1L
SDP
Catalog No. NC1026538
Encompass_Preferred
The stop solution provides minimal signal drift compared to sulfuric or hydrochloric acid. Absorbance is read in the yellow spectral range (450 nm) and can be read up to 3 hours after the TMB reaction has been stopped using the Nova-Stop Solution.
Catalog No. NC1026538 Supplier Surmodics Inc Supplier No. NSTP100001
The stop solution provides minimal signal drift compared to sulfuric or hydrochloric acid. Absorbance is read in the yellow spectral range (450 nm) and can be read up to 3 hours after the TMB reaction has been stopped using the Nova-Stop Solution.