Learn More
Surmodics Inc 450 nm Stop Reagent for TMB Microwell Substrates, 1L
SDP
Catalog No. NC0213329
Encompass_Preferred
A dry blend stop reagent used in end point ELISA applications where a TMB substrate is used as the detection mechanism. Absorbance is read in the yellow spectral range (450 nm).
Catalog No. NC0213329 Supplier Surmodics Inc Supplier No. STPR100001
A dry blend stop reagent used in end point ELISA applications where a TMB substrate is used as the detection mechanism. Absorbance is read in the yellow spectral range (450 nm).