Learn More
ABclonal Technology 450 Rabbit anti-Human CD116
SDP
Catalog No. 5000008516
Encompass_Preferred
The protein encoded by this gene is the alpha subunit of the heterodimeric receptor for colony stimulating factor 2 a cytokine which controls the production differentiation and function of granulocytes and macrophages The encoded protein is a member of the cytokine family of receptors This gene is found in the pseudoautosomal region (PAR) of the X and Y chromosomes Multiple transcript variants encoding different isoforms have been found for this gene with some of the isoforms being membrane-bound and others being soluble
Catalog No. 50-000-08516 Supplier ABclonal Technology Supplier No. A26805200T
The protein encoded by this gene is the alpha subunit of the heterodimeric receptor for colony stimulating factor 2 a cytokine which controls the production differentiation and function of granulocytes and macrophages The encoded protein is a member of the cytokine family of receptors This gene is found in the pseudoautosomal region (PAR) of the X and Y chromosomes Multiple transcript variants encoding different isoforms have been found for this gene with some of the isoforms being membrane-bound and others being soluble