Learn More
ABclonal Technology 450 Rabbit anti-Human CD2
Supplier: ABclonal Technology A26937500T
The protein encoded by this gene is a surface antigen found on all peripheral blood T-cells The encoded protein interacts with LFA3 (CD58) on antigen presenting cells to optimize immune recognition A locus control region (LCR) has been found in the 3 flanking sequence of this gene