Learn More
ABclonal Technology 450 Rabbit anti-Human CD66b
SDP
Catalog No. 5000010825
Encompass_Preferred
Enables protein heterodimerization activity Involved in heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules Located in cell surface and extracellular space Biomarker of severe acute respiratory syndrome
Catalog No. 50-000-10825 Supplier ABclonal Technology Supplier No. A26461100T
Enables protein heterodimerization activity Involved in heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules Located in cell surface and extracellular space Biomarker of severe acute respiratory syndrome