Learn More
ABclonal Technology 450 Rabbit anti-Human CD97
SDP
Catalog No. 5000008864
Encompass_Preferred
This gene encodes a member of the EGF-TM7 subfamily of adhesion G protein-coupled receptors which mediate cell-cell interactions These proteins are cleaved by self-catalytic proteolysis into a large extracellular subunit and seven-span transmembrane subunit which associate at the cell surface as a receptor complex The encoded protein may play a role in cell adhesion as well as leukocyte recruitment activation and migration and contains multiple extracellular EGF-like repeats which mediate binding to chondroitin sulfate and the cell surface complement regulatory protein CD55 Expression of this gene may play a role in the progression of several types of cancer Alternatively spliced transcript variants encoding multiple isoforms with 3 to 5 EGF-like repeats have been observed for this gene This gene is found in a cluster with other EGF-TM7 genes on the short arm of chromosome 19
Catalog No. 50-000-08864 Supplier ABclonal Technology Supplier No. A26361500T
This gene encodes a member of the EGF-TM7 subfamily of adhesion G protein-coupled receptors which mediate cell-cell interactions These proteins are cleaved by self-catalytic proteolysis into a large extracellular subunit and seven-span transmembrane subunit which associate at the cell surface as a receptor complex The encoded protein may play a role in cell adhesion as well as leukocyte recruitment activation and migration and contains multiple extracellular EGF-like repeats which mediate binding to chondroitin sulfate and the cell surface complement regulatory protein CD55 Expression of this gene may play a role in the progression of several types of cancer Alternatively spliced transcript variants encoding multiple isoforms with 3 to 5 EGF-like repeats have been observed for this gene This gene is found in a cluster with other EGF-TM7 genes on the short arm of chromosome 19