Learn More
ABclonal Technology 450 Rabbit anti-Human IgD
SDP
Catalog No. 5000011347
Encompass_Preferred
IgD is the major antigen receptor isotype on the surface of most peripheral B-cells where it is coexpressed with IgM The membrane-bound IgD (mIgD) induces the phosphorylation of CD79A and CD79B by the Src family of protein tyrosine kinases
Catalog No. 50-000-11347 Supplier ABclonal Technology Supplier No. A26480200T
IgD is the major antigen receptor isotype on the surface of most peripheral B-cells where it is coexpressed with IgM The membrane-bound IgD (mIgD) induces the phosphorylation of CD79A and CD79B by the Src family of protein tyrosine kinases