Learn More
Sigma Aldrich Fine Chemicals Biosciences 450nm Light Source1EA
Catalog No. 502908914
Encompass
The 450nm LED Light Module is used with the Penn PhD Photoreactor M2 for chemists and researchers to accelerate chemical reactions using photoredox catalysis.Features and Benefits of Penn Penn PhD Photoreactor M2Modular design allows for use with a variety of wavelengths 365nm - 450nm360 degree reflective environment maximizes surface area photon captureLight shield interlock prevents user exposure to harmful light raysInteractive touch screen controls reaction parametersIntertek ETL CE and CB approvedUser defined parameters including temperature light intensity fan speed and stirringAuto stop pause and reset optionsSupports vial sizes gc 4 8 20 40 mlTemp feedback using a k-type thermocouplePhotocatalysis Technology Spotlight
Catalog No. 50-290-8914 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7440331EA
The 450nm LED Light Module is used with the Penn PhD Photoreactor M2 for chemists and researchers to accelerate chemical reactions using photoredox catalysis.Features and Benefits of Penn Penn PhD Photoreactor M2Modular design allows for use with a variety of wavelengths 365nm - 450nm360 degree reflective environment maximizes surface area photon captureLight shield interlock prevents user exposure to harmful light raysInteractive touch screen controls reaction parametersIntertek ETL CE and CB approvedUser defined parameters including temperature light intensity fan speed and stirringAuto stop pause and reset optionsSupports vial sizes gc 4 8 20 40 mlTemp feedback using a k-type thermocouplePhotocatalysis Technology Spotlight