Learn More
Surmodics Inc 450 nm Liquid Stop Solution for TMB Microwell Substrates, 100mL
SDP
Catalog No. NC9427003
Encompass_Preferred
450 nm Liquid Stop Solution for TMB Microwell Substrates (100 mL) is a proprietary liquid used to stop TMB microwell substrate reactions causing a color change from blue to yellow. Absorbance is read in the yellow spectral range (450 nm).
Catalog No. NC9427003 Supplier Surmodics Inc Supplier No. LSTP010001
450 nm Liquid Stop Solution for TMB Microwell Substrates (100 mL) is a proprietary liquid used to stop TMB microwell substrate reactions causing a color change from blue to yellow. Absorbance is read in the yellow spectral range (450 nm).