Learn More
Surmodics Inc 450 nm Stop Reagent for TMB Microwell Substrates, 100mL
SDP
Catalog No. NC9797912
Encompass_Preferred
450 nm Stop Reagent for TMB Microwell Substrates (100 mL) is a proprietary dry blend used to stop TMB microwell substrate reactions causing a color change from blue to yellow. The yellow color is read in at 450 nm.
Catalog No. NC9797912 Supplier Surmodics Inc Supplier No. STPR010001
450 nm Stop Reagent for TMB Microwell Substrates (100 mL) is a proprietary dry blend used to stop TMB microwell substrate reactions causing a color change from blue to yellow. The yellow color is read in at 450 nm.