Learn More
Ace Glass, Inc. UV lamp, immersion, 450 watt, 121.92mm arc length, 244.35mmoverall length, metal-ended, no insulation, NEMA 15P power cord included
SDP
Catalog No. 509558006
Encompass
450watt med. pressure, quartz mercury-vapor lamp, for use in all ACE immersion wells. Arc length: 121.92mm. Overall length: 244.35mm. Supplied with 61cm PTFE covered lead wires, fitted with pin type connectors, and 6 cord to power supply.
Catalog No. 50-955-8006 Supplier Ace Glass, Inc. Supplier No. 782534
450watt med. pressure, quartz mercury-vapor lamp, for use in all ACE immersion wells. Arc length: 121.92mm. Overall length: 244.35mm. Supplied with 61cm PTFE covered lead wires, fitted with pin type connectors, and 6 cord to power supply.