Learn More
DEROYAL MEDICAL PRODUCTS 4511 RED HANDLE SCALPEL 50/CS
Catalog No. 50371503CS
Encompass_Preferred
#11 Safety Scalpel, Blue Slider Retractable Safety Scalpel 50/CS. Meets US FDA requirements for the use of sharps with engineered safety protection. Easy to operate and now with permanent lock for final disposal. Not made with natural rubber latex. Packed sterile.
This item is not returnable. View return policy
Catalog No. 50371503CS Supplier DEROYAL MEDICAL PRODUCTS Supplier No. D4511
#11 Safety Scalpel, Blue Slider Retractable Safety Scalpel 50/CS. Meets US FDA requirements for the use of sharps with engineered safety protection. Easy to operate and now with permanent lock for final disposal. Not made with natural rubber latex. Packed sterile.