Learn More
Cayman Chemical 4567 ttrabromobenzIMDazo 25mg
SDP
Catalog No. 502982446
Encompass_Preferred
A selective, ATP-competitive inhibitor of CK2 with Ki values ranging from 0.5-1 µM
Catalog No. 50-298-2446 Supplier Cayman Chemical Supplier No. 1888625 MG
A selective, ATP-competitive inhibitor of CK2 with Ki values ranging from 0.5-1 µM