Learn More
Alkali Scientific 45mL Flip Top Polypropylene Sample Vials (2oz.), 200/Pk, 600/Case
SDP
Catalog No. 5000661962
Encompass_Preferred
45mL Flip Top Polypropylene Sample Vials from Alkali Scientific provide a reliable solution for the collection, transport, storage, and packaging of liquid and solid samples. Designed for routine laboratory and field applications, these vials offer dependable performance without locking or tamper evident features. Manufactured from food grade polypropylene, they provide excellent durability and chemical resistance for food and beverage sampling, environmental and water testing, dairy analysis, veterinary use, and general laboratory workflows. The airtight, leak resistant construction helps protect sample integrity during handling and transport. A permanent hinged flip top lid enables convenient one handed operation while preventing cap loss and reducing contamination risk, while molded graduations provide a clear, permanent volume reference.
Catalog No. 50-006-61962 Supplier Alkali Scientific Supplier No. FTSV45
45mL Flip Top Polypropylene Sample Vials from Alkali Scientific provide a reliable solution for the collection, transport, storage, and packaging of liquid and solid samples. Designed for routine laboratory and field applications, these vials offer dependable performance without locking or tamper evident features. Manufactured from food grade polypropylene, they provide excellent durability and chemical resistance for food and beverage sampling, environmental and water testing, dairy analysis, veterinary use, and general laboratory workflows. The airtight, leak resistant construction helps protect sample integrity during handling and transport. A permanent hinged flip top lid enables convenient one handed operation while preventing cap loss and reducing contamination risk, while molded graduations provide a clear, permanent volume reference.