Learn More
Bellco Glass, Inc. 45mm Complete/HOSE BARB Cap Assembly 4-45 Bioreactor Flask
SDP
Catalog No. 501945029
Encompass_Preferred
Bioreactor Hose Barb AssemblyThe Stainless Steel 45mm compression and hose barb cap assemblyis designed with a 1/4” ID compression fitting for insertion of tube accessories and 1/4” OD hose barb for silicone tubing attachment. The compression fitting is supplied with a nylon ferrule for easy adjustment of tubes. A bluepolypropylene open top cap and white silicone gasket are included. MADE IN THE USA. --316L stainless steel compression fitting and hose barb--Blue polypropylene 45mm screw cap with open top--White silicone rubber gasket
Catalog No. 50-194-5029 Supplier Bellco Glass, Inc. Supplier No. 196545200
Bioreactor Hose Barb AssemblyThe Stainless Steel 45mm compression and hose barb cap assemblyis designed with a 1/4” ID compression fitting for insertion of tube accessories and 1/4” OD hose barb for silicone tubing attachment. The compression fitting is supplied with a nylon ferrule for easy adjustment of tubes. A bluepolypropylene open top cap and white silicone gasket are included. MADE IN THE USA. --316L stainless steel compression fitting and hose barb--Blue polypropylene 45mm screw cap with open top--White silicone rubber gasket