Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cascade Columbia Distribution Potassium Hydroxide 45% w/w solution, 3750 lb Tote, clear, colorless, odorless liquid
SDP

Catalog No. NC2939344
Encompass_Preferred

This product is a highly concentrated 45% w/w solution of Potassium Hydroxide (KOH), commonly known as Caustic Potash. It is produced via the electrolysis of potassium chloride and is essential for various industrial applications due to its strong alkaline properties. Safety protocols are crucial when handling this corrosive solution, especially during dilution due to exothermic reactions.

  • Clear, colorless and odorless liquid
  • Food Grade (FCC)
  • Density: 1.46 g/mL at 20-25 °C
  • pH >12, highly alkaline
  • UN Number: UN1814 (corrosive)
  • Molecular Formula: KOH
  • Boiling Point: 132.2 °C (270 °F)
  • Freeze/Melting Point: -28.8 °C (-20 °F)
  • Completely soluble in water
  • Key applications include pH adjustment, saponification, and chemical synthesis

Catalog No. NC2939344 Supplier Cascade Columbia Distribution Supplier No. 242000TT
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This product is a highly concentrated 45% w/w solution of Potassium Hydroxide (KOH), commonly known as Caustic Potash. It is produced via the electrolysis of potassium chloride and is essential for various industrial applications due to its strong alkaline properties. Safety protocols are crucial when handling this corrosive solution, especially during dilution due to exothermic reactions.

  • Clear, colorless and odorless liquid
  • Food Grade (FCC)
  • Density: 1.46 g/mL at 20-25 °C
  • pH >12, highly alkaline
  • UN Number: UN1814 (corrosive)
  • Molecular Formula: KOH
  • Boiling Point: 132.2 °C (270 °F)
  • Freeze/Melting Point: -28.8 °C (-20 °F)
  • Completely soluble in water
  • Key applications include pH adjustment, saponification, and chemical synthesis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.