Learn More
Chemglass Life Sciences #45 Loosening Ring
SDP
Catalog No. 501531934
Encompass_Preferred
#45 Loosening Ring. Split Polyamide loosening ring to enable inner and outer Rodaviss members to be pulled apart in the event of freezing.
Catalog No. 50-153-1934 Supplier Chemglass Life Sciences Supplier No. CGB18406
#45 Loosening Ring. Split Polyamide loosening ring to enable inner and outer Rodaviss members to be pulled apart in the event of freezing.