Learn More
Waters Corp Cuvette Kit, 45 μL, Quartz, for NanoStar, NanoStar II, Mobius or ZetaStar
Catalog No. NC1810879 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit includes a 45 μL Quartz Cuvette and essential cleaning accessories, ensuring optimal performance and longevity. It is designed for seamless integration with a range of laboratory instruments.
  • Includes a 45 μL Quartz Cuvette
  • Comes with lens tissue and foam applicators for cleaning
  • Compatible with NanoStar, NanoStar II, Mobius, and ZetaStar systems
  • Requires a cuvette adapter for use with Mobius
Catalog No. NC1810879 Supplier Waters Corp Supplier No. WNQC4500
This kit includes a 45 μL Quartz Cuvette and essential cleaning accessories, ensuring optimal performance and longevity. It is designed for seamless integration with a range of laboratory instruments.
  • Includes a 45 μL Quartz Cuvette
  • Comes with lens tissue and foam applicators for cleaning
  • Compatible with NanoStar, NanoStar II, Mobius, and ZetaStar systems
  • Requires a cuvette adapter for use with Mobius