Learn More
Labsciences LabSciences OFT6006 Dissolution Filter Tip For +/- 1/8 dia Cannula/Pipettes|1000
Catalog No. NC9314251
Encompass_Preferred
An economical alternative for cannula and pipette filters. Ultra high molecular weight (UHMW) polyethylene with porosities ranging from 2 to 90 micron. Designed to fit most sampling cannula and pipettes.
Catalog No. NC9314251 Supplier Labsciences Supplier No. OFT6006
An economical alternative for cannula and pipette filters. Ultra high molecular weight (UHMW) polyethylene with porosities ranging from 2 to 90 micron. Designed to fit most sampling cannula and pipettes.