Learn More
Medchemexpress LLC HY-N7433 50mg Medchemexpress, 4,6-O-Ethylidene-α-D-glucose CAS:13224-99-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN743350MG
Encompass_Preferred
Medchemexpress, HY-N7433 50mg 4,6-O-Ethylidene-α-D-glucose CAS:13224-99-2 4,6-O-ethylidene-α-D-glucose (Ethylidene-glucose), a glucose derivative, is a competitive exofacial binding-site inhibitor on glucose transporter 1 (GLUT1) with a Ki of 12 mM for wild-type 2-deoxy-D-glucose transport. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2997