Learn More
MSC Clear 3M Double-Sided Acrylic Foam Tape, 27 yd
Catalog No. 501298679
Encompass
This is a clear, continuous roll of double-sided acrylic foam tape designed for strong, long-lasting adhesion in various applications. It offers clean removability and good temperature resistance.
  • Clear, 0.031 in (0.8 mm) closed-cell acrylic foam carrier
  • Clean removability from many substrates
  • High ultimate adhesion to a wide variety of surfaces
  • Unique acrylic adhesive system provides long term durability
  • Good initial and long-term temperature resistance
Catalog No. 50-129-8679 Supplier MSC Supplier No. 00188888
This is a clear, continuous roll of double-sided acrylic foam tape designed for strong, long-lasting adhesion in various applications. It offers clean removability and good temperature resistance.
  • Clear, 0.031 in (0.8 mm) closed-cell acrylic foam carrier
  • Clean removability from many substrates
  • High ultimate adhesion to a wide variety of surfaces
  • Unique acrylic adhesive system provides long term durability
  • Good initial and long-term temperature resistance