Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UltraSep™ 47 mm Polyethersulfone Disc

Catalog No. 09924046 Shop All GVS Products
Change view
Click to view available options
Pore Size:
0.45 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
09-924-046 0.45 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-924-046 Supplier UltraSep™ Supplier No. 1214475
Add to Cart
Add to Cart

Constructed from a high-temperature polyethersulfone polymer that is acid and base resistant. Its strength and durability are advantageous during usage that involves aggressive handling or automated equipment. It's naturally hydrophilic w/o added wetting agents and has low extractables.

Properties

  • Hydrophilic: eliminates the need for wetting agents that can potentially interfere with analyses.
  • Low extractables: ensures test results will not be compromised by wetting agents or other extractables.
  • Low drug and protein binding: maximizes recovery of critical drugs or proteins.
  • Wide range of pore sizes: pore size range of 0.03 μm to 5.0 μm enables specific pore size selection for given applications.
  • Superior burst strength: protects the integrity of the membrane under high pressure.
  • Quality checks ensure lot-to-lot consistency, both down and across the polyester web, for dependable results every time.

Characteristics

  • Pore size: 0.03 μm to 5.0 μm
  • Thickness: 110 μm to 150 μm
  • Membrane material: PES

Applications

  • Biological fluid sterilization
  • Environmental water studies
  • Pharmaceutical sterilization
  • Protein and enzyme filtration

Specifications

Autoclavable Autoclavable
Disposable Limited-use
Filter Type Membrane Filter
Format Disc
Material PES
Pore Size 0.45 μm
Sterility Non-sterile
Type Disc Membrane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.