Learn More
Aviva Systems Biology Corp 4732418C07Rik Antibody - C-terminal region (ARP47074_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47074_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against 4732418C07Rik. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of 4732418C07Rik. Alternate names: KIAA0494, Kiaa0494, RP23-146I18.2, 4732418C07RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-286