Learn More
Aviva Systems Biology Corp 4732471D19Rik Antibody - C-terminal region (ARP55911_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55911_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against 4732471D19Rik. It was validated on Western Blot. . Alternate names: 4732471D19Rik, ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-096