Learn More
Andwin Scientific 47MM INLINE FILTER HOLDER
SDP
Supplier:  Andwin Scientific 1119
Encompass_Preferred
The 47 mm In-line Filter Holder has a lightweight plastic construction. This product is ideal for venting and in-line liquid or gas filtration at either indoor or outdoor locations. The filter holder has a body constructed with polycarbonate. It has a diameter of 47 mm. This is sold individually.
Catalog No. NC9843897