Learn More
Cytiva (Formerly Pall Lab) 47 mm In-Line Filter Holder, Stainless Steel - 47 mm stainless steel (1/pkg)
Supplier:  Cytiva 2220
Encompass
High-quality, corrosion-resistant in-line filter holder is designed for stack sampling.Membrane or glass fiber filtersHolder opens and closes easilyFilter remains undisturbed for particulate analysisUpstream threaded vent allows release of trapped gas during liquid filtrationBody, center ring collar, support screen, and underdrain disc are made of Type 316 stainless steel. Captive thrust ring is PTFE; O-ring is Viton rubber; hose barb adapters are polyethylene. Inlet/outlet connections: 9.5 mm (3/8") FNPT. Hose barbs accept 6.4 mm (1/4") I.D. tubing. Autoclavable. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1347267