Learn More
Ace Glass, Inc. Fritted disc, 47mm OD, porosity B (70-100 micron), fits3702-10
SDP
Catalog No. 50978189
Encompass_Preferred
47mm O.D. Porosity B (70-100 micron) fritted disc.
Catalog No. 50-978-189 Supplier Ace Glass, Inc. Supplier No. 370347
47mm O.D. Porosity B (70-100 micron) fritted disc.