Learn More
C&G Containers 48-400 Open-Top Cap With PTFE-Lined Septum, Secure Seal, Ideal For Labs, 100 Units
SDP
Catalog No. NC1421760
Encompass_Preferred
The 48-400 open-top cap with PTFE-lined septum is ideal for laboratory applications that require sample integrity and secure sealing. This closure, manufactured by C & G CONTAINERS, facilitates easy needle penetration and offers chemical resistance, making it suitable for high-volume operations. With a packaging quantity of 100 units, it stands out for its reliability in preventing contamination during analysis.
Catalog No. NC1421760 Supplier C&G Containers Supplier No. CAH481000I04
The 48-400 open-top cap with PTFE-lined septum is ideal for laboratory applications that require sample integrity and secure sealing. This closure, manufactured by C & G CONTAINERS, facilitates easy needle penetration and offers chemical resistance, making it suitable for high-volume operations. With a packaging quantity of 100 units, it stands out for its reliability in preventing contamination during analysis.