Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ 48-400 Polypropylene Caps, Unlined
SDP

Catalog No. 02927141 Shop All DWK Life Sciences Products
Catalog No. 02-927-141 Supplier DWK Life Sciences Supplier No. W240543
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WHEATON™ 48-400 Polypropylene Caps, Unlined are white, unlined polypropylene caps compatible with WHEATON™ screw cap bottles and vials. Featuring a 48mm outer diameter and GSI 400 thread finish, sold in cases of 1700.

  • For use with Wheaton screw cap bottles and vials
  • Opening outer diameter (including threads): 48mm
  • Thread finish: GSI 400 (1 turn)
  • Material: White polypropylene
  • Unlined, no liner insert
  • Sold in cases of 1700
TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
Quantity 1700/Case
Sterility Non-sterile
Neck Finish 48-400
For Use With (Equipment) Wheaton screw cap bottles/vials
Autoclavable Autoclavable
Disposable Reusable
Product Type Unlined Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.