Learn More
O Berk Company Plastic CT Closure: 48-400 black rib side/matte top pp plastic screw cap with F217 liner
Catalog No. NC1710298
Encompass_Preferred
Plastic CT Closure: 48-400 black rib side/matte top pp plastic screw cap with F217 liner
Catalog No. NC1710298 Supplier O Berk Company Supplier No. 48400OV
Plastic CT Closure: 48-400 black rib side/matte top pp plastic screw cap with F217 liner