Learn More
CELLTREAT Scientific Products 4.6mL 48 Deep Well Storage Plate, PP, Rectangle Well, U-Bottom, Non-sterile
SDP
Catalog No. NC1964628
Encompass_Preferred
48 Well Deep Storage Plate features larger, rectangular well for increased well volume. Square to round transition of V-Bottom maximizes working volume. Pyramid Bottom eases sample retrieval and improves mixing by creating a sloshing effect rather than a vortex when agitated. Round Bottom with round wells enable use with magnetic beads. Polypropylene offers a low binding surface to prevent samples from sticking to the side walls during elution, and is chemically inert for combinatorial chemistry applications.
Catalog No. NC1964628 Supplier CELLTREAT Scientific Products Supplier No. 229578
48 Well Deep Storage Plate features larger, rectangular well for increased well volume. Square to round transition of V-Bottom maximizes working volume. Pyramid Bottom eases sample retrieval and improves mixing by creating a sloshing effect rather than a vortex when agitated. Round Bottom with round wells enable use with magnetic beads. Polypropylene offers a low binding surface to prevent samples from sticking to the side walls during elution, and is chemically inert for combinatorial chemistry applications.