Learn More
Analytical Sales & Services 48 Hole Kapton Plate (1mil Liner) with Chamfered Corners
Catalog No. 5000805691
Encompass
48 Hole Stainless Steel Plate with Adhesive-Bonded Kapton Film Liner\n• Liner thickness:  1mil\n• SS plate thickness: 1.5mm\n• Well diameter: 8.8mm\n• Used to collect Powder X-Ray Diffraction data on polymorph screening samples\n• Designed for both transmission and/or reflection instruments\n\nQty/Pkg: 10
Catalog No. 50-008-05691 Supplier Analytical Sales & Services Supplier No. 4857710222
48 Hole Stainless Steel Plate with Adhesive-Bonded Kapton Film Liner\n• Liner thickness:  1mil\n• SS plate thickness: 1.5mm\n• Well diameter: 8.8mm\n• Used to collect Powder X-Ray Diffraction data on polymorph screening samples\n• Designed for both transmission and/or reflection instruments\n\nQty/Pkg: 10