Learn More
Uline Cold Packs - 48 oz,1 EA
Catalog No. NC0463621
Encompass_Preferred
Reusable leakproof refrigerant gel stays colder than ice. Freeze before use. Dependable, durable and reusable. Thick 4 mil poly. FDA compliant. Solid when frozen, not flexible. Gel is non-toxic, food safe. Can be used to prevent freezing if heated in a microwave. Heat on low setting for 30 second increments. Flip pack over every 30 seconds until warm. Outer bag can be recycled. Cut open to remove the gel. Gel can be disposed of in garbage. Do not place down the drain.
Catalog No. NC0463621 Supplier Uline Supplier No. S12762
Reusable leakproof refrigerant gel stays colder than ice. Freeze before use. Dependable, durable and reusable. Thick 4 mil poly. FDA compliant. Solid when frozen, not flexible. Gel is non-toxic, food safe. Can be used to prevent freezing if heated in a microwave. Heat on low setting for 30 second increments. Flip pack over every 30 seconds until warm. Outer bag can be recycled. Cut open to remove the gel. Gel can be disposed of in garbage. Do not place down the drain.