Learn More
Research Products International Corp Micro-Plate Pin Replicator, Flat Square Design, 1.5mm x 1.5mm Pin Size, 48 Pin
SDP
Catalog No. 50212190
Encompass
Micro-Plate Pin Replicators transfer small volume samples to plates, membranes or solid supports. Ideal for inoculum, clones, YACs, cosmid libraries, colony hybridization, antibiotic sensitivity testing, phage typing, replication of DNA libraries and more. Individual pins transfer approximately 1µl of sample. Transfer volume may vary depending on amount of pressure applied and viscosity of sample.
Pins are made of stainless steel for durability, autoclaving and flame sterilization. An economical substitute for expensive robotics systems.
Catalog No. 50-212-190 Supplier Research Products International Corp Supplier No. 140501
Micro-Plate Pin Replicators transfer small volume samples to plates, membranes or solid supports. Ideal for inoculum, clones, YACs, cosmid libraries, colony hybridization, antibiotic sensitivity testing, phage typing, replication of DNA libraries and more. Individual pins transfer approximately 1µl of sample. Transfer volume may vary depending on amount of pressure applied and viscosity of sample.
Pins are made of stainless steel for durability, autoclaving and flame sterilization. An economical substitute for expensive robotics systems.