Learn More
Thomson Instrument Company Multi-Use Press - 48 Position for Thomson Filter Vials
SDP
Catalog No. 501120014
Encompass_Preferred
The Thomson Filter Vial Press enables high solid content and viscous liquids to be easily filtered through vials. Some fermentation cultures that reach 100 OD or particulate laden samples may require the toggle press. Filter vial presses are available in 5 and 48-vial configurations. The press allows for consistent ergonomic use of filter vials. The 48-position block can be transitioned to some automation and autosampler systems directly. Thomson manufactures its products in the USA to assure same day shipping.
Catalog No. 50-112-0014 Supplier Thomson Instrument Company Supplier No. 35015
The Thomson Filter Vial Press enables high solid content and viscous liquids to be easily filtered through vials. Some fermentation cultures that reach 100 OD or particulate laden samples may require the toggle press. Filter vial presses are available in 5 and 48-vial configurations. The press allows for consistent ergonomic use of filter vials. The 48-position block can be transitioned to some automation and autosampler systems directly. Thomson manufactures its products in the USA to assure same day shipping.